Anti-PCDHB6 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP12006A
89380-338EA 580.97 USD
89380-338
Anti-PCDHB6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PCDHB6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR