Sodium octadecyl sulfate ≥98% (dry weight)

Synonyms: Octadecyl sulphuric acid saltSodium Stearyl sulphateSodium Stearyl sulfateSodium n-octadecylsulphaten-Octadecyl sulphate, sodium saltOctadecyl sulfuric acid saltn-Octadecyl sulfate, sodium saltSodium n-octadecylsulfate

225620-5G 225620-25G
BT225620-5GEA 217.25 USD
BT225620-5G BT225620-25G
Sodium octadecyl sulfate ≥98% (dry weight)
Sodium octadecyl sulphate

  • RTECS: WT1170000
  • Powder

Formula: C₁₈H₃₇NaO₄S
MW: 372.55 g/mol
Melting Pt: 211…213 °C
MDL Number: MFCD00007469
CAS Number: 1120-04-3
EINECS: 214-295-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR