Boc-N-methyl-D-alanine ≥98%
Supplier: BeanTown Chemical
Synonyms:
Boc-N-Me-Ala-OH
, N-Boc-N-methyl-L-alanine
224130-1G
224130-5G
BT224130-1GEA
88.91
USD
BT224130-1G
BT224130-5G
Boc-N-methyl-D-alanine ≥98%
Boc-N-methyl-D-alanine
- Optical rotation: [α]20/D -30±1°, c = 1% in ethanol
- Powder
Formula:
C₉H₁₇NO₄ MW: 203.2 g/mol |
MDL Number:
MFCD00037242 CAS Number: 16948-16-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...