Morpholine hydrochloride

Supplier: BeanTown Chemical

Synonyms: Morpholinium chloride

224075-5G 224075-1G
BT224075-1GEA 129.39 USD
BT224075-1G BT224075-5G
Morpholine hydrochloride
Morpholine hydrochloride

  • Solid

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD00035287
CAS Number: 10024-89-2
EINECS: 233-029-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR