Methyl-2-bromohexanoate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Methyl 2-bromohexanoate
223295-25ML
223295-100ML
BT223295-25MLEA
39.85
USD
BT223295-25ML
BT223295-100ML
Methyl-2-bromohexanoate ≥99%
Methyl 2-bromohexanoate
- Refractive index: 1.455
- Liquid
Formula:
C₇H₁₃BrO₂ MW: 209.08 g/mol Boiling Pt: 82 °C/60 mmHg Density: 1.289 g/mL Flash Pt: 69 °C (156 °F) |
EINECS:
226-643-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...