cis-1,2,3,6-Tetrahydrophthalic anhydride ≥95%

Supplier: BeanTown Chemical

Synonyms: cis-4-Cyclohexene-1,2-dicarboxylic anhydride

222565-250G
BT222565-250GEA 24.83 USD
BT222565-250G
cis-1,2,3,6-Tetrahydrophthalic anhydride ≥95%
cis-1,2,3,6-Tetrahydrophthalic anhydride

  • RTECS: GW5775000
  • Moisture Sensitive
  • Flakes

Formula: C₈H₈O₃
Melting Pt: 97…103 °C
Flash Pt: 156 °C (312 °F)
MDL Number: MFCD00005916
CAS Number: 935-79-5
EINECS: 213-308-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR