6-tert-Butyl-2,4-xylenol ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-(tert-Butyl)-4,6-dimethylphenol

222370-25G
BT222370-25GEA 32 USD
BT222370-25G
6-tert-Butyl-2,4-xylenol ≥97%
6-tert-Butyl-2,4-xylenol

  • Liquid/Low Melting Solid

Formula: C₁₂H₁₈O
MW: 178.28 g/mol
Boiling Pt: 249 °C
Melting Pt: 22…23 °C
Density: 0.917 g/mL
MDL Number: MFCD00002234
EINECS: 217-533-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR