β-Alanine ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Aminopropanoic acidH-β-Ala-OHH-Bala-OH3-Aminopropionic acid

221795-250G 221795-5KG 221795-1KG
BT221795-1KGEA 77.91 USD
BT221795-1KG BT221795-250G BT221795-5KG
β-Alanine ≥98%
β-Alanine

  • RTECS: UA2369200
  • Crystalline

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Melting Pt: 202 °C (dec.)
Density: 1.437 g/mL
MDL Number: MFCD00008200
CAS Number: 107-95-9
EINECS: 203-536-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR