Methylammonium chloride ≥99%

Synonyms: Methylamine hydrochlorideAminomethane hydrochlorideMethylammonium chlorideMACl

221660-2.5KG
BT221660-2.5KGEA 279.4 USD
BT221660-2.5KG
Methylammonium chloride ≥99%
Methylammonium chloride

  • RTECS: PA0603000
  • Hygroscopic
  • Crystalline

Formula: CH₆ClN
MW: 67.52 g/mol
Boiling Pt: 225…230 °C/15 mmHg
Melting Pt: 231…233 °C
MDL Number: MFCD00012849
CAS Number: 593-51-1
EINECS: 209-795-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR