6(5H)-Phenanthridinone ≥96%

Supplier: BeanTown Chemical

221490-1G 221490-5G
BT221490-1GEA 43.84 USD
BT221490-1G BT221490-5G
6(5H)-Phenanthridinone ≥96%
6(5H)-Phenanthridinone

  • RTECS: SG0370000
  • Solid

Formula: C₁₃H₉NO
MW: 195.22 g/mol
Melting Pt: 290…292 °C
MDL Number: MFCD00004988
CAS Number: 1015-89-0
EINECS: 213-804-3

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR