Lithium sulfate, anhydrous ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
221470-500G
221470-100G
221470-25G
BT221470-25GEA
129.39
USD
BT221470-25G
BT221470-500G
BT221470-100G
Lithium sulfate, anhydrous ≥99.99% (trace metals basis)
Lithium sulphate
- Hygroscopic
- Powder
Formula:
Li₂O₄S MW: 109.95 g/mol Melting Pt: 845 °C Density: 2.22 g/mL |
MDL Number:
MFCD00011086 CAS Number: 10377-48-7 EINECS: 233-820-4 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...