BEAN220770

220770-25G 220770-100G
BT220770-25GEA 50.11 USD
BT220770-25G BT220770-100G
BEAN220770
CAS: 21732-17-2; EC No: 244-551-7; MDL No: MFCD00038813
Solid; Molecular Formula: C3H3N4O2; MW: 127.08
Melting Point: 127-129°
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR