N,N'-Diphenylguanidine monohydrobromide

Supplier: BeanTown Chemical

Synonyms: N,N-Diphenylguanidine Monohydrobromide

220670-5G
BT220670-5GEA 49.44 USD
BT220670-5G
N,N'-Diphenylguanidine monohydrobromide
N,N'-Diphenylguanidine monohydrobromide

  • Solid

Formula: C₁₃H₁₄BrN₃
MW: 292.18 g/mol
MDL Number: MFCD04038163
CAS Number: 93982-96-8
EINECS: 301-283-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR