Calcium iodide, anhydrous ≥99.5%

Supplier: BeanTown Chemical

220645-25G 220645-100G
BT220645-25GEA 77.51 USD
BT220645-25G BT220645-100G
Calcium iodide, anhydrous ≥99.5%
Calcium iodide

CAS: 10102-68-8; EC No: 233-276-8; MDL No: MFCD00010910
-20 Mesh Powder; Molecular Formula: CaI2; MW: 293.89
Melting Point: 779°
Hygroscopic , Light Sensitive

Formula: CaI₂
MW: 293,89 g/mol
Boiling Pt: 1100 °C (1013 hPa)
Melting Pt: 740 °C
Density: 3.956 g/cm³ (25 °C)
MDL Number: MFCD00010910
CAS Number: 10102-68-8
EINECS: 233-276-8
Merck Index: 12,01720

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR