1,4,7,10-Tetraazacyclododecane ≥99%

Supplier: BeanTown Chemical

220560-250MG
BT220560-250MGEA 146.41 USD
BT220560-250MG
1,4,7,10-Tetraazacyclododecane ≥99%
1,4,7,10-Tetraazacyclododecane (Cyclen, Kryptofix® 11 aza)

  • RTECS: XA5253000
  • Hygroscopic
  • Crystalline

Formula: C₈H₂₀N₄
MW: 172.27 g/mol
Melting Pt: 110…113 °C
MDL Number: MFCD00066281
EINECS: 202-928-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR