1-(3-Cyanopropyl)pyrrolidine ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Pyrrolidinebutyronitrile1-Pyrrolidinebutanenitrile4-Pyrrolidinobutyronitrile

220280-25G
BT220280-25GEA 34.68 USD
BT220280-25G
1-(3-Cyanopropyl)pyrrolidine ≥97%
1-(3-Cyanopropyl)pyrrolidine

  • RTECS: UY0270000
  • Refractive index: 1.46
  • Liquid

Formula: C₈H₁₄N₂
MW: 138.21 g/mol
Boiling Pt: 115 °C/18 mmHg
Density: 0.926 g/mL
Flash Pt: 99 °C (210 °F)
MDL Number: MFCD00003183
CAS Number: 35543-25-0
EINECS: 252-612-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR