5-Bromotryptamine hydrochloride ≥98%
Supplier: BeanTown Chemical
220185-250MG
220185-1G
BT220185-250MGEA
97.12
USD
BT220185-250MG
BT220185-1G
5-Bromotryptamine hydrochloride ≥98%
5-Bromotryptamine hydrochloride
- Light Sensitive
- Crystalline/Powder
Formula:
C₁₀H₁₂BrClN₂ Melting Pt: 275 °C (dec.) |
MDL Number:
MFCD00152022 CAS Number: 81868-12-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...