Salbutamol sulfate ≥99%

Supplier: BeanTown Chemical

Synonyms: Albutarol sulphateBronchosprayProventil-HFA2-(tert-Butylamino)-1-(4-hydroxy-3-hydroxymethylphenyl)ethanol sulphateAlbuterol hemisulfateAlbutarol sulfate2-(tert-Butylamino)-1-(4-hydroxy-3-hydroxymethylphenyl)ethanol sulfateAlbuterol hemissulphateSalbutamol hemisulphate5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzyl Alcohol sulphateSalbutamol hemisulfate5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzyl Alcohol sulfate

219960-1G 219960-25G 219960-5G
BT219960-1GEA 56.91 USD
BT219960-1G BT219960-5G BT219960-25G
Salbutamol sulfate ≥99%
Salbutamol sulphate

  • RTECS: ZE4400000
  • Powder

Formula: (C₁₃H₂₁NO₃)₂·H₂SO₄
MW: 576.702 g/mol
MDL Number: MFCD00055200
CAS Number: 51022-70-9
EINECS: 242-424-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR