5-Fluoro-4-hydrazinyl-2-methoxypyrimidine ≥95%

Supplier: BeanTown Chemical

219840-1G
BT219840-1GEA 815.37 USD
BT219840-1G
5-Fluoro-4-hydrazinyl-2-methoxypyrimidine ≥95%
5-Fluoro-4-hydrazinyl-2-methoxypyrimidine

  • Solid

Formula: C₅H₇FN₄O
CAS Number: 166524-64-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR