Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥97%

Supplier: BeanTown Chemical

Synonyms: N-Hydroxysulfosuccinimide sodium saltsulfo-N-HydroxysuccinimideHydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium saltN-Hydroxy sulfosuccinimide sodium saltSulfo NHSN-Hydroxysulphosuccinimide sodium saltHydroxy-2,5-dioxopyrrolidine-3-sulfonic acid sodium saltN-Hydroxy sulphosuccinimide sodium saltSulpho-N-Hydroxysuccinimide

219395-1G 219395-5G 219395-250MG
BT219395-1GEA 271.09 USD
BT219395-1G BT219395-250MG BT219395-5G
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥97%
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt)

  • Moisture Sensitive
  • Powder

Formula: C₄H₄NNaO₆S
MW: 217.13 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR