Hafnium(IV) tert-butoxide ≥99.9% (trace metals basis excluding Zr)

Supplier: BeanTown Chemical

Synonyms: Hafnium(IV) t-butoxideHafnium(IV)-tert-butanolate

219065-1G 219065-5G
BT219065-1GEA 80.3 USD
BT219065-1G BT219065-5G
Hafnium(IV) tert-butoxide ≥99.9% (trace metals basis excluding Zr)
Hafnium(IV) tert-butoxide

  • Refractive index: 1.424
  • Moisture Sensitive
  • Liquid

Formula: C₁₆H₃₆HfO₄
MW: 470.94 g/mol
Boiling Pt: 90 °C/5 mmHg
Density: 1.166 g/mL
Flash Pt: 28 °C (82 °F)
MDL Number: MFCD00070458
CAS Number: 2172-02-3

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR