Saponin

Supplier: BeanTown Chemical

219060-100G 219060-25G
BT219060-100GEA 101.43 USD InStock
BT219060-100G BT219060-25G
Saponin
Saponin

CAS: 8047-15-2; EC No: 232-462-6; MDL No: MFCD00081981; RTECS: VQ1400000
Solid
Melting Point: 158° (decomposes)
Density (g/mL): 1.015

Density: 1.015 to 1.02 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00081981
CAS Number: 8047-15-2
EINECS: 232-462-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR