DL-Threonine ≥99%
Supplier: BeanTown Chemical
219010-100G
219010-500G
219010-25G
BT219010-25GEA
32
USD
BT219010-25G
BT219010-500G
BT219010-100G
DL-Threonine ≥99%
DL-Threonine
- Powder
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 244 °C (dec.) |
MDL Number:
MFCD00063722 CAS Number: 80-68-2 EINECS: 201-300-6 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...