n-Triacontane ≥99%
Supplier: BeanTown Chemical
218990-5G
218990-1G
218990-250MG
BT218990-250MGEA
22.34
USD
BT218990-250MG
BT218990-5G
BT218990-1G
n-Triacontane ≥99%
n-Triacontane
- Flakes
Formula:
H₃C(CH₂)₂₈CH₃ MW: 422.82 g/mol Boiling Pt: 455 °C Melting Pt: 64…67 °C Flash Pt: 237 °C (458 °F) |
MDL Number:
MFCD00009410 CAS Number: 638-68-6 EINECS: 211-349-5 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...