4-Methoxyphenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: (4-Methoxyphenyl)metaboric acidDihydroxy(4-methoxyphenyl)borane4-Methoxybenzeneboronic acid

218220-1G 218220-25G 218220-5G
BT218220-1GEA 29.85 USD InStock
BT218220-1G BT218220-25G BT218220-5G
4-Methoxyphenylboronic acid ≥97%
4-Methoxyphenylboronic acid

CAS: 5720-07-0; MDL No: MFCD00039139; RTECS: CY8975000
Powder; Molecular Formula: C7H9BO3; MW: 151.96
Melting Point: 204-206°

Formula: C₇H₉BO₃
MW: 151,96 g/mol
Melting Pt: 204 to 207 °C
Storage Temperature: ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0
EINECS: 000-000-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR