5-Nitroguaiacol sodium salt ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Methoxy-5-nitrophenol Sodium Salt

218175-25G
BT218175-25GEA 163.04 USD
BT218175-25G
5-Nitroguaiacol sodium salt ≥98%
5-Nitroguaiacol sodium salt

  • Solid

Formula: C₇H₇NO₄
MW: 191.12 g/mol
Boiling Pt: 291 °C
Melting Pt: 105…106 °C
MDL Number: MFCD00070570
CAS Number: 67233-85-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR