Iodosobenzene diacetate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Iodobenzene diacetate
218135-100G
218135-25G
218135-500G
BT218135-25GEA
66.71
USD
BT218135-25G
BT218135-100G
BT218135-500G
Iodosobenzene diacetate ≥98%
Iodosobenzene diacetate
- RTECS: DA3525000
- Light Sensitive
- Powder
Formula:
C₁₀H₁₁IO₄ MW: 322.1 g/mol Melting Pt: 161…163 °C |
MDL Number:
MFCD00008692 CAS Number: 3240-34-4 EINECS: 221-808-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...