BEAN218005

218005-100G 218005-25G 218005-500G
BT218005-500GEA 353.31 USD
BT218005-500G BT218005-25G BT218005-100G
BEAN218005
CAS: 69-93-2; EC No: 200-720-7; MDL No: MFCD00005712; RTECS: YU7050080
Crystalline; Molecular Formula: C5H4N4O3; MW: 168.11
Melting Point: 300°
Density (g/mL): 1.89
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR