(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥97%

Supplier: BeanTown Chemical

Synonyms: Benzyl (S)-Tetrahydro-5-oxo-3-furanylcarbamate(S)-beta-(Cbz-amino)-gamma-butyrolactone(S)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one(S)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate(S)-4-(Cbz-amino)tetrahydrofuran-2-one

217965-5G
BT217965-5GEA 206.67 USD
BT217965-5G
(S)-β-(Carbobenzoxyamino)-γ-butyrolactone ≥97%
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone

  • Optical rotation: [α]21/D -49°, c = 1 in acetone
  • Powder

Formula: C₁₂H₁₃NO₄
MW: 235.24 g/mol
Melting Pt: 103…105 °C
MDL Number: MFCD00216569
CAS Number: 87219-29-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR