Sodium aurothiomalate ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Sodium aurothiomalate(I)

217830-1G 217830-5G
BT217830-5GEA 1408.49 USD
BT217830-5G BT217830-1G
Sodium aurothiomalate ≥99.9% (trace metals basis)
Sodium aurothiomalate

  • RTECS: MD5435000
  • Hygroscopic
  • Powder

Formula: C₄H₆AuNaO₄S
MW: 370,11 g/mol
MDL Number: MFCD00064304
CAS Number: 12244-57-4
EINECS: 235-479-7

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR