3-Nitrophthalhydrazide ≥98%

Synonyms: 3-Nitrophthalic Hydrazide5-Nitro-2,3-dihydrophthalazine-1,4-dione2,3-Dihydro-5-nitro-1,4-phthalazinedione3-Nitrophthalhydrazide

217375-25G
BT217375-25GEA 151.49 USD
BT217375-25G
3-Nitrophthalhydrazide ≥98%
3-Nitrophthalhydrazide

  • Powder

Formula: C₈H₅N₃O₄
MW: 207.1430 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00006889
CAS Number: 3682-15-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR