(3-Bromo-4-methoxyphenyl)acetic acid

Supplier: BeanTown Chemical

Synonyms: 3-Bromo-4-methoxyphenylacetic acid2-(3-Bromo-4-methoxyphenyl)acetic acid

217085-5G 217085-1G
BT217085-5GEA 279.4 USD
BT217085-5G BT217085-1G
(3-Bromo-4-methoxyphenyl)acetic acid
(3-Bromo-4-methoxyphenyl)acetic acid

  • Solid

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD00017538
CAS Number: 774-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR