D-(+)-Maltose 20% in water

Synonyms: (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal

216760-100ML
BT216760-100MLEA 81.59 USD
BT216760-100ML
D-(+)-Maltose 20% in water
D(+)-Maltose

  • Refractive index: 1.361
  • Liquid

Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Melting Pt: 110 °C
Density: 1.070 g/mL
MDL Number: MFCD00135877
CAS Number: 69-79-4
EINECS: 200-716-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR