3-Coumaranone ≥97%

Supplier: BeanTown Chemical

Synonyms: Benzofuran-3(2H)-one

216380-25G 216380-5G 216380-1G
BT216380-5GEA 169.15 USD
BT216380-5G BT216380-25G BT216380-1G
3-Coumaranone ≥97%
3-Coumaranone

  • Light Sensitive
  • Crystalline

Formula: C₈H₆O₂
MW: 134.13
Melting Pt: 101…103 °C
MDL Number: MFCD00051810
CAS Number: 7169-34-8
EINECS: 000-000-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR