1,3-Diethylurea ≥97%

Supplier: BeanTown Chemical

Synonyms: N,N'-Diethylurea

216335-5G 216335-25G
BT216335-5GEA 27.01 USD
BT216335-5G BT216335-25G
1,3-Diethylurea ≥97%
1,3-Diethylurea

  • RTECS: YS9354000
  • Powder

Formula: C₅H₁₂N₂O
Melting Pt: 109…113 °C
MDL Number: MFCD00009028
CAS Number: 623-76-7
EINECS: 210-811-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR