1-Dodecanethiol ≥98%

Supplier: BeanTown Chemical

Synonyms: n-Dodecyl mercaptan1-Mercaptododecanen-Dodecanethiol

215975-500G 215975-100G
BT215975-100GEA 26.81 USD
BT215975-100G BT215975-500G
1-Dodecanethiol ≥98%
1-Dodecanethiol

  • RTECS: JR3155000
  • Refractive index: 1.459
  • Air Sensitive
  • Liquid

Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 266…283 °C
Density: 0.845 g/mL
Flash Pt: 120 °C (248 °F)
MDL Number: MFCD00004885
CAS Number: 112-55-0
EINECS: 203-984-1
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR