Gallium(III) ethanolate

Synonyms: Gallium(III) ethoxide

215755-25G 215755-1G
BT215755-25GEA 1301.13 USD
BT215755-25G BT215755-1G
Gallium(III) ethanolate
Gallium(III) ethanolate

  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₅GaO₃
MW: 204.91 g/mol
Flash Pt: 68 °C (154 °F)
MDL Number: MFCD01866730
CAS Number: 2572-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR