Bis[2-(diphenylphosphino)phenyl]ether ≥98%
Supplier: BeanTown Chemical
Synonyms:
Bis(2-diphenylphosphinophenyl)ether
, (Oxybis(2,1-phenylene))bis(diphenylphosphine)
, 2,2'-Bis(diphenylphosphino)diphenyl Ether
, DPEphos
, (Oxydi-2,1-phenylene)bis(diphenylphosphine)
215095-1G
215095-5G
BT215095-5GEA
155.8
USD
BT215095-5G
BT215095-1G
Bis[2-(diphenylphosphino)phenyl]ether ≥98%
Bis[2-(diphenylphosphino)phenyl]ether
- Solid
Formula:
C₃₆H₂₈OP₂ MW: 538.57 g/mol Melting Pt: 184…187 °C |
MDL Number:
MFCD00233863 CAS Number: 166330-10-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...