Pentaethylene glycol ≥98%

Supplier: BeanTown Chemical

Synonyms: 3,6,9,12-Tetraoxatetradecane-1,14-diol

213205-10G 213205-250G 213205-50G
BT213205-50GEA 388.44 USD
BT213205-50G BT213205-10G BT213205-250G
Pentaethylene glycol ≥98%
Pentaethylene glycol

  • RTECS: RZ2670000
  • Refractive index: 1.462
  • Hygroscopic
  • Liquid

Formula: C₁₀H₂₂O₆
MW: 238.28 g/mol
Boiling Pt: 338…340 °C
Density: 1.126 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002878
CAS Number: 4792-15-8
EINECS: 225-341-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR