1,3-Phenyldiboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,3-Benzenediboronic acid1,3-Phenyldiboric acidm-Phenylenebisboronic acid1,3-Phenylenediboronic acid

212675-1G 212675-25G 212675-5G
BT212675-1GEA 151.18 USD
BT212675-1G BT212675-25G BT212675-5G
1,3-Phenyldiboronic acid ≥97%
1,3-Phenyldiboronic acid

  • Solid

Formula: C₆H₈B₂O₄
MW: 165.75 g/mol
MDL Number: MFCD03092915
CAS Number: 4612-28-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR