Luminol (3-Aminophthalhydrazide) ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Amino-2,3-dihydrophthalazine-1,4-dioneNSC 50645-Amino-2,3-Dihydro-1,4-Phthalazinedione

212320-100G 212320-25G 212320-5G
BT212320-5GEA 48.31 USD
BT212320-5G BT212320-100G BT212320-25G
Luminol (3-Aminophthalhydrazide) ≥98%
Luminol (3-Aminophthalhydrazide)

  • RTECS: TH8890060
  • Powder

Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00006890
CAS Number: 521-31-3
EINECS: 208-309-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR