4-Vinylphenylboronic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: (4-Vinylphenyl)metaboric acidDihydroxy(4-vinylphenyl)borane4-Vinylbenzeneboronic acid

211650-1G 211650-25G 211650-5G
BT211650-5GEA 112.15 USD
BT211650-5G BT211650-25G BT211650-1G
4-Vinylphenylboronic acid ≥97%
4-Vinylphenylboronic acid

  • Solid

Formula: C₈H₉BO₂
MW: 147.97 g/mol
Melting Pt: 190…193 °C
MDL Number: MFCD00239441
CAS Number: 2156-04-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR