12-Hydroxystearic acid ≥95%

Supplier: BeanTown Chemical

Synonyms: 12-Hydroxyoctadecanoic Acid

211455-1G 211455-25G 211455-5G
BT211455-1GEA 22.24 USD
BT211455-1G BT211455-25G BT211455-5G
12-Hydroxystearic acid ≥95%
12-Hydroxystearic acid

  • RTECS: W13700000
  • Crystalline

Formula: C₁₈H₃₆O₃
MW: 300.48 g/mol
Melting Pt: 74…76 °C
MDL Number: MFCD00004592
CAS Number: 106-14-9
EINECS: 203-366-1

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR