2,3-Dihydro-1H-carbazol-4(9H)-one ≥95%
Supplier: BeanTown Chemical
Synonyms:
1,2,3,4-Tetrahydro-4-oxocarbazole
, 1,2,3,4-Tetrahydrocarbazol-4-one
211400-25G
211400-5G
BT211400-25GEA
229.54
USD
BT211400-25G
BT211400-5G
2,3-Dihydro-1H-carbazol-4(9H)-one ≥95%
2,3-Dihydro-1H-carbazol-4(9H)-one
- Solid
Formula:
C₁₂H₁₁NO MW: 185.2218 g/mol Boiling Pt: 386 °C Melting Pt: 224…226 °C |
MDL Number:
MFCD00173749 CAS Number: 15128-52-6 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...