Kieselguhr, powder -325 Mesh

Supplier: BeanTown Chemical

Synonyms: Siliceous earthDiatomiteInfusorial earthDiatomaceous silicaCeliteDiatomaceous earth

211300-5KG 211300-1KG
BT211300-1KGEA 41.48 USD
BT211300-1KG BT211300-5KG
Kieselguhr, powder -325 Mesh
Kieselguhr

  • RTECS: EU8655000

Formula: O₂Si
MW: 60.08 g/mol
MDL Number: MFCD00132803
CAS Number: 7631-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR