Perfluorohexane ≥98% (85% n-isomer)

Supplier: BeanTown Chemical

Synonyms: Tetradecafluorohexane

211170-100G 211170-25G 211170-500G
BT211170-500GEA 237.85 USD
BT211170-500G BT211170-25G BT211170-100G
Perfluorohexane ≥98% (85% n-isomer)
Perfluorohexane

  • RTECS: MO4310000
  • Refractive index: 1.252
  • Liquid

Formula: C₆F₁₄
MW: 338.04 g/mol
Boiling Pt: 58…60 °C
Melting Pt: –4 °C
Density: 1.669 g/mL
MDL Number: MFCD00000437
CAS Number: 355-42-0
EINECS: 206-585-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR