L-Threonine ≥98%
Supplier: BeanTown Chemical
Synonyms:
(2S,3R)-(-)-Threonine
, (2S,3R)-2-Amino-3-hydroxybutyric acid
, H-Thr-OH
211050-500G
211050-100G
211050-25G
BT211050-25GEA
54.93
USD
BT211050-25G
BT211050-500G
BT211050-100G
L-Threonine ≥98%
L-Threonine
- RTECS: XO8590000
- Crystalline/Powder
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 255 °C (dec.) |
MDL Number:
MFCD00064270 CAS Number: 72-19-5 EINECS: 200-774-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...