Croconic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,5-Dihydroxycyclopent-4-ene-1,2,3-trione4,5-Dihydroxy-4-cyclopentene-1,2,3-trione

145855-5G 145855-1G
BT145855-5GEA 693.85 USD
BT145855-5G BT145855-1G
Croconic acid ≥98%
Croconic acid

  • Powder

Formula: C₅H₂O₅
MW: 142.07 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00181389
CAS Number: 488-86-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR