1,3,5-Triazine ≥97%

Supplier: BeanTown Chemical

Synonyms: s-Triazine

145550-1G 145550-5G 145550-25G
BT145550-1GEA 38.84 USD
BT145550-1G BT145550-25G BT145550-5G
1,3,5-Triazine ≥97%
1,3,5-Triazine

  • RTECS: XY2957000
  • Moisture Sensitive
  • Crystalline

Formula: C₃H₃N₃
MW: 81.08 g/mol
Melting Pt: 77…83 °C (dec.)
MDL Number: MFCD00006044
CAS Number: 290-87-9
EINECS: 206-028-1
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR