Calcium iodide, anhydrous ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical


Ampuled under argon

145195-25G 145195-500G 145195-100G
BT145195-25GEA 111.79 USD InStock
BT145195-25G BT145195-500G BT145195-100G
Calcium iodide, anhydrous ≥99.99% (trace metals basis)
Calcium iodide

CAS: 10102-68-8; EC No: 233-276-8; MDL No: MFCD00010910
Powder; Molecular Formula: CaI2 ; MW: 293.89
Melting Point: 779°
Hygroscopic , Light Sensitive

Formula: CaI₂
MW: 293,89 g/mol
Boiling Pt: 1100 °C (1013 hPa)
Melting Pt: 740 °C
Density: 3.956 g/cm³ (25 °C)
MDL Number: MFCD00010910
CAS Number: 10102-68-8
EINECS: 233-276-8
Merck Index: 12,01720

Order Now

Specification Test Results

Assay 99.99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR